Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Deoxycholic acid | 83-44-3 | 392.57 | 1 ML
SDP

Catalog No. 5000388705
Encompass_Preferred

Deoxycholic acid is a bile acid and a by-product of intestinal metabolism. It activates the G protein-coupled bile acid receptor TGR5. This product is intended for research use only.

  • Activates G protein-coupled bile acid receptor TGR5
  • High purity of 99.94%
  • Appearance as a white to off-white solid
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO (≥ 100 mg/mL)

Catalog No. 50-003-88705 Supplier Medchemexpress LLC Supplier No. HYN059310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Deoxycholic acid is a bile acid and a by-product of intestinal metabolism. It activates the G protein-coupled bile acid receptor TGR5. This product is intended for research use only.

  • Activates G protein-coupled bile acid receptor TGR5
  • High purity of 99.94%
  • Appearance as a white to off-white solid
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO (≥ 100 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.