Learn More
Medchemexpress LLC Deoxycholic acid | 83-44-3 | 392.57 | 5 G
SDP
Catalog No. 5000388785
Encompass_Preferred
Deoxycholic acid (cholanoic acid) is a bile acid, a by-product of intestinal metabolism that activates the G protein-coupled bile acid receptor TGR5. It is for research use only.
  • Activates G protein-coupled bile acid receptor TGR5
  • Available in solid form
  • Suitable for research applications
Catalog No. 50-003-88785 Supplier Medchemexpress LLC Supplier No. HYN05935G
Deoxycholic acid (cholanoic acid) is a bile acid, a by-product of intestinal metabolism that activates the G protein-coupled bile acid receptor TGR5. It is for research use only.
  • Activates G protein-coupled bile acid receptor TGR5
  • Available in solid form
  • Suitable for research applications