Learn More
Cayman Chemical Deoxycholic Acid-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 112076-61-6, 10 mg
SDP
Catalog No. 502985567
Encompass_Preferred
An internal standard for the quantification of DCA by GC- or LC-MS
This item is not returnable. View return policy
Catalog No. 50-298-5567 Supplier Cayman Chemical Supplier No. 2085110 MG
An internal standard for the quantification of DCA by GC- or LC-MS