Learn More
Medchemexpress LLC Deoxycholic acid-d6 5mg
SDP
Catalog No. 5000669396
Encompass_Preferred
Deoxycholic acid-d6 is the deuterium labeled Deoxycholic acid Deoxycholic acid is specifically responsible for activating the G protein-coupled bile acid receptor TGR5 that stimulates brown adipose tissue (BAT) thermogenic activity
Catalog No. 50-006-69396 Supplier Medchemexpress LLC Supplier No. 5MGHYN0593S2
Deoxycholic acid-d6 is the deuterium labeled Deoxycholic acid Deoxycholic acid is specifically responsible for activating the G protein-coupled bile acid receptor TGR5 that stimulates brown adipose tissue (BAT) thermogenic activity