Learn More
Crescent Chemical Co Inc DEOXYCHOLIC ACID-NA-SALT PURE
SDP
Catalog No. 50247474
Encompass_Preferred
Deoxycholic acid-Na-salt pure For bacteriology and enzymology. Suitable for solubilization of many membrane proteins and phospholipids. Size - 100G Storage Conditions - +15 °C TO +30 °C Catalog Number - 18330.03 CAS 302-95-4
Catalog No. 50-247-474 Supplier Crescent Chemical Co Inc Supplier No. SE18330.03
Deoxycholic acid-Na-salt pure For bacteriology and enzymology. Suitable for solubilization of many membrane proteins and phospholipids. Size - 100G Storage Conditions - +15 °C TO +30 °C Catalog Number - 18330.03 CAS 302-95-4