Learn More
Crescent Chemical Co Inc DEOXYCHOLIC ACID-NA-SALT PURE
SDP
Supplier:  Crescent Chemical Co Inc SE18330.02
Encompass_Preferred
Deoxycholic acid-Na-salt pure For bacteriology and enzymology. Suitable for solubilization of many membrane proteins and phospholipids. Size - 25G Storage Conditions - +15 °C TO +30 °C Catalog Number - 18330.02 CAS 302-95-4
Catalog No. 50-247-473