Learn More
Medchemexpress LLC Deoxycholic acid sod 10mg
SDP
Catalog No. 5000669534
Encompass_Preferred
Deoxycholic acid sodium salt (Standard) is the analytical standard of Deoxycholic acid sodium salt This product is intended for research and analytical applications Deoxycholic acid sodium salt (sodium deoxycholate) a bile acid is a by-product of intestinal metabolism that activates the G protein-coupled bile acid receptorTGR5[1][2]
Catalog No. 50-006-69534 Supplier Medchemexpress LLC Supplier No. 10MGHYN0593AR
Deoxycholic acid sodium salt (Standard) is the analytical standard of Deoxycholic acid sodium salt This product is intended for research and analytical applications Deoxycholic acid sodium salt (sodium deoxycholate) a bile acid is a by-product of intestinal metabolism that activates the G protein-coupled bile acid receptorTGR5[1][2]