Learn More
Bioworld Deoxycholic Acid, Sodium Sal, 100 g
Catalog No. 50255876
Encompass_Preferred
Deoxycholic Acid, Sodium SaltSynonyms: Sodium Deoxycholate, 3α,12α-Dihydroxy-5β-cholanic acid sodium salt, 7-Deoxycholic acid sodium saltAn ionic detergent for the solubilization of membrane-bound proteinsHigh Purity GradeHumidity < 0.4%RTECS Number: FZ2250000
Catalog No. 50-255-876 Supplier Bioworld Supplier No. 404300042
Deoxycholic Acid, Sodium SaltSynonyms: Sodium Deoxycholate, 3α,12α-Dihydroxy-5β-cholanic acid sodium salt, 7-Deoxycholic acid sodium saltAn ionic detergent for the solubilization of membrane-bound proteinsHigh Purity GradeHumidity < 0.4%RTECS Number: FZ2250000