Learn More
Selleck Chemical LLC Deoxycholic acid sodium salt S3069-25mg
SDP
Catalog No. 503111580
Encompass_Preferred
Deoxycholic acid sodium salt (Sodium Deoxycholate Sodium Desoxycholate Cholanoic Acid ATX-101 Kythera Belkyra DCA) is specifically responsible for activating the TGR5 receptor (GPCR19) that stimulates brown adipose tissue (BAT) thermogenic activity
Catalog No. 50-311-1580 Supplier Selleck Chemical LLC Supplier No. S306925MG
Deoxycholic acid sodium salt (Sodium Deoxycholate Sodium Desoxycholate Cholanoic Acid ATX-101 Kythera Belkyra DCA) is specifically responsible for activating the TGR5 receptor (GPCR19) that stimulates brown adipose tissue (BAT) thermogenic activity