Learn More
Medchemexpress LLC Deoxycholic acid sodium salt | 302-95-4 | 414.55 | 500 MG
SDP
Catalog No. 5000388652
Encompass_Preferred
Deoxycholic acid sodium salt is a bile acid, a by-product of intestinal metabolism, that activates the G protein-coupled bile acid receptor TGR5. It is for research use only and not sold to patients.
  • Activates G protein-coupled bile acid receptor TGR5
  • High purity (99.89%)
  • White to off-white solid appearance
  • Suitable for research in intestinal metabolism and biological activity studies
  • Stable at 4°C for solid form, and in solvent at -80°C or -20°C for extended periods
Catalog No. 50-003-88652 Supplier Medchemexpress LLC Supplier No. HYN0593A500MG
Deoxycholic acid sodium salt is a bile acid, a by-product of intestinal metabolism, that activates the G protein-coupled bile acid receptor TGR5. It is for research use only and not sold to patients.
  • Activates G protein-coupled bile acid receptor TGR5
  • High purity (99.89%)
  • White to off-white solid appearance
  • Suitable for research in intestinal metabolism and biological activity studies
  • Stable at 4°C for solid form, and in solvent at -80°C or -20°C for extended periods