Learn More
Abcam Deo x ycholic acid sodium salt, Ionic detergent, 50G
Catalog No. 502704740
Encompass_Preferred
MW 414.56 g/mol. Water-soluble ionic detergent useful for extraction of membrane receptors and other plasma membrane proteins. Reported to induce tyrosine phosphorylation of β-catenin and its subsequent translocation to the nucleus, promoting activation of β-catenin signaling (effective concentration 5 - 50 μM).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4740 Supplier Abcam Supplier No. AB14630550G
MW 414.56 g/mol. Water-soluble ionic detergent useful for extraction of membrane receptors and other plasma membrane proteins. Reported to induce tyrosine phosphorylation of β-catenin and its subsequent translocation to the nucleus, promoting activation of β-catenin signaling (effective concentration 5 - 50 μM).
The product is subject to the following: Abcam Restricted Use Statement