Learn More
Medchemexpress LLC Deoxycholic acid (Standard) | 83-44-3 | 392.57 | 50 MG
SDP
Catalog No. 5000388927
Encompass_Preferred
Deoxycholic acid (Standard) is an analytical standard intended for research and analytical applications. It is also known as cholanoic acid, a bile acid that is a by-product of intestinal metabolism and activates the G protein-coupled bile acid receptor TGR5.
  • Used in qualitative research experiments such as HPLC, GC, and MS.
  • Used in quantitative research experiments such as HPLC, GC, and MS.
  • Used in methodological research experiments such as HPLC, GC, and MS.
Catalog No. 50-003-88927 Supplier Medchemexpress LLC Supplier No. HYN0593R50MG
Deoxycholic acid (Standard) is an analytical standard intended for research and analytical applications. It is also known as cholanoic acid, a bile acid that is a by-product of intestinal metabolism and activates the G protein-coupled bile acid receptor TGR5.
  • Used in qualitative research experiments such as HPLC, GC, and MS.
  • Used in quantitative research experiments such as HPLC, GC, and MS.
  • Used in methodological research experiments such as HPLC, GC, and MS.