Learn More
Cayman Chemical Deoxycorticosterone Acetate, Purity: ≥95%, CAS: 56-47-3, 10 g
SDP
Catalog No. 502989346
Encompass_Preferred
A corticosteroid; has commonly been used in rodents as a model of salt-sensitive hypertension, characterized by increases in sympathetic nervous system activity, imbalances in the renin-angiotensin system (RAS), and cardiac remodeling in combination with saline
Catalog No. 50-298-9346 Supplier Cayman Chemical Supplier No. 3085610 G
A corticosteroid; has commonly been used in rodents as a model of salt-sensitive hypertension, characterized by increases in sympathetic nervous system activity, imbalances in the renin-angiotensin system (RAS), and cardiac remodeling in combination with saline