Learn More
Medchemexpress LLC HY-113414 10mg Medchemexpress, Deoxycorticosterone CAS:64-85-7 Purity:>98%
SDP
Catalog No. 501969253
Encompass_Preferred
Medchemexpress, HY-113414 10mg Deoxycorticosterone CAS:64-85-7 Deoxycorticosterone is a steroid hormone produced by the adrenal gland that possesses mineralocorticoid activity and acts as an aldosterone precursor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9253 Supplier Medchemexpress LLC Supplier No. HY11341410MG
Medchemexpress, HY-113414 10mg Deoxycorticosterone CAS:64-85-7 Deoxycorticosterone is a steroid hormone produced by the adrenal gland that possesses mineralocorticoid activity and acts as an aldosterone precursor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.