Learn More
Selleck Chemical LLC Deoxycorticosterone acetate S4243-50mg
SDP
Catalog No. 503109088
Encompass_Preferred
Deoxycorticosterone acetate (11-Deoxycorticosterone DOC Cortexone) is a steroid hormone used for intramuscular injection for replacement therapy of the adrenocortical steroid Deoxycorticosterone acetate can be used to induce animal models of Aneurysm Hypertension
Catalog No. 50-310-9088 Supplier Selleck Chemical LLC Supplier No. S424350MG
Deoxycorticosterone acetate (11-Deoxycorticosterone DOC Cortexone) is a steroid hormone used for intramuscular injection for replacement therapy of the adrenocortical steroid Deoxycorticosterone acetate can be used to induce animal models of Aneurysm Hypertension