Learn More
Selleck Chemical LLC Deoxycorticosterone acetate S4243-1g
SDP
Catalog No. 503111116
Encompass_Preferred
Deoxycorticosterone acetate (11-Deoxycorticosterone DOC Cortexone) is a steroid hormone used for intramuscular injection for replacement therapy of the adrenocortical steroid Deoxycorticosterone acetate can be used to induce animal models of Aneurysm Hypertension
Catalog No. 50-311-1116 Supplier Selleck Chemical LLC Supplier No. S42431G
Deoxycorticosterone acetate (11-Deoxycorticosterone DOC Cortexone) is a steroid hormone used for intramuscular injection for replacement therapy of the adrenocortical steroid Deoxycorticosterone acetate can be used to induce animal models of Aneurysm Hypertension