Learn More
Medchemexpress LLC Deoxycytidine 5'-triphosphate, trisodium salt | 109909-44-6 | 99.5% | 533.10 g/mol | C9H13N3Na3O13P3 | 25 MG
SDP
Catalog No. 5000211154
Encompass_Preferred
Deoxycytidine triphosphate (trisodium) is provided as a ready-to-use 100 mM aqueous solution of the trisodium salt of dCTP. It functions as a nucleotide substrate for DNA synthesis, polymerase reactions, and enzymatic assays, and is supplied as a high-purity solution intended for molecular biology workflows.
  • High purity: 99.46% suitable for sensitive assays.
  • Ready-to-use 100 mM aqueous solution for immediate use.
  • Trisodium salt form enhances solubility in aqueous buffers.
  • Colorless to light yellow liquid for easy visual inspection.
  • Available in small pack sizes (for example, 25 mg) to reduce waste.
  • Storage-stable when frozen; follow manufacturer storage guidance.
Catalog No. 50-002-11154 Supplier Medchemexpress LLC Supplier No. HY101400A25MG
Deoxycytidine triphosphate (trisodium) is provided as a ready-to-use 100 mM aqueous solution of the trisodium salt of dCTP. It functions as a nucleotide substrate for DNA synthesis, polymerase reactions, and enzymatic assays, and is supplied as a high-purity solution intended for molecular biology workflows.
  • High purity: 99.46% suitable for sensitive assays.
  • Ready-to-use 100 mM aqueous solution for immediate use.
  • Trisodium salt form enhances solubility in aqueous buffers.
  • Colorless to light yellow liquid for easy visual inspection.
  • Available in small pack sizes (for example, 25 mg) to reduce waste.
  • Storage-stable when frozen; follow manufacturer storage guidance.