Learn More
Medchemexpress LLC Deoxycytidine triphosphate-15N3 (dilithium) | 2056-98-6 | MFCD00135938 | 99.0% | 482.00 g·mol⁻1 | C9H14[15N]3Li2O13P3 | 1 MG
SDP
Catalog No. 5000211147
Encompass_Preferred
Deoxycytidine triphosphate-15N3 (dilithium) is a stable isotope-labeled deoxynucleoside triphosphate provided as a concentrated aqueous solution for molecular biology and analytical applications. The 15N3 label enables sensitive and specific detection in mass spectrometry and tracer experiments, while the dilithium salt formulation offers good aqueous solubility. Typical uses include use as an internal standard, tracer, or reagent in DNA synthesis and sequencing workflows.
  • Supplied as a concentrated solution for convenient dilution and use.
  • 15N3 isotopic label supports quantitative mass spectrometry and tracer studies.
  • High chemical purity for reliable analytical and preparative performance.
  • Suitable as an internal standard or tracer in nucleic acid workflows.
  • Compatible with PCR, cDNA synthesis, and DNA sequencing applications.
  • Dilithium salt form improves solubility in aqueous buffers.
Catalog No. 50-002-11147 Supplier Medchemexpress LLC Supplier No. HY101400S31MG
Deoxycytidine triphosphate-15N3 (dilithium) is a stable isotope-labeled deoxynucleoside triphosphate provided as a concentrated aqueous solution for molecular biology and analytical applications. The 15N3 label enables sensitive and specific detection in mass spectrometry and tracer experiments, while the dilithium salt formulation offers good aqueous solubility. Typical uses include use as an internal standard, tracer, or reagent in DNA synthesis and sequencing workflows.
  • Supplied as a concentrated solution for convenient dilution and use.
  • 15N3 isotopic label supports quantitative mass spectrometry and tracer studies.
  • High chemical purity for reliable analytical and preparative performance.
  • Suitable as an internal standard or tracer in nucleic acid workflows.
  • Compatible with PCR, cDNA synthesis, and DNA sequencing applications.
  • Dilithium salt form improves solubility in aqueous buffers.