Learn More
Medchemexpress LLC Deoxycytidine triphosphate | 2056-98-6 | MFCD00135938 | 98.2% | 467.16 g/mol | C9H16N3O13P3 | 50 MG
SDP
Catalog No. 5000211153
Encompass_Preferred
Deoxycytidine triphosphate (dCTP) is a deoxynucleoside triphosphate used as a substrate for DNA polymerases in molecular biology. Supplied as a laboratory reagent, it is intended for enzymatic DNA synthesis applications such as PCR, cDNA synthesis, and DNA sequencing.
  • High purity suitable for enzymatic DNA synthesis and PCR.
  • Accurate molecular weight and formula for reagent planning.
  • Supplied in a convenient small-scale package for method development.
  • Stable when stored under recommended conditions to preserve activity.
  • Commonly used as a deoxynucleotide substrate in sequencing and amplification workflows.
Catalog No. 50-002-11153 Supplier Medchemexpress LLC Supplier No. HY10140050MG
Add to Cart
Add to Cart
Deoxycytidine triphosphate (dCTP) is a deoxynucleoside triphosphate used as a substrate for DNA polymerases in molecular biology. Supplied as a laboratory reagent, it is intended for enzymatic DNA synthesis applications such as PCR, cDNA synthesis, and DNA sequencing.
  • High purity suitable for enzymatic DNA synthesis and PCR.
  • Accurate molecular weight and formula for reagent planning.
  • Supplied in a convenient small-scale package for method development.
  • Stable when stored under recommended conditions to preserve activity.
  • Commonly used as a deoxynucleotide substrate in sequencing and amplification workflows.