Learn More
Medchemexpress LLC Deoxycytidine triphosphate-d14 dilithium | 2687960-70-7 | 95.7% | 493.11 g/mol | C9D14Li2N3O13P3 | 10 MG
SDP
Catalog No. 5000211143
Encompass_Preferred
Deoxycytidine triphosphate-d14 dilithium is a deuterium-labeled nucleoside triphosphate provided as the dilithium salt for use as an isotopic standard or tracer in DNA synthesis workflows, including PCR, sequencing, and analytical method development.
  • Deuterium-labeled for isotopic tracing and mass-shift detection.
  • Dilithium salt form, compatible with aqueous buffers.
  • High purity (~95.7%) suitable for analytical and molecular biology use.
  • Supplied in a 10 mg vial for small-scale experiments.
  • Useful as a stable labeled standard for qPCR, sequencing, and method validation.
Catalog No. 50-002-11143 Supplier Medchemexpress LLC Supplier No. HY101400S10MG
Deoxycytidine triphosphate-d14 dilithium is a deuterium-labeled nucleoside triphosphate provided as the dilithium salt for use as an isotopic standard or tracer in DNA synthesis workflows, including PCR, sequencing, and analytical method development.
  • Deuterium-labeled for isotopic tracing and mass-shift detection.
  • Dilithium salt form, compatible with aqueous buffers.
  • High purity (~95.7%) suitable for analytical and molecular biology use.
  • Supplied in a 10 mg vial for small-scale experiments.
  • Useful as a stable labeled standard for qPCR, sequencing, and method validation.