Learn More
Medchemexpress LLC Deoxycytidine triphosphate disodium | 102783-51-7 | 99.0% | 511.12 | 5 MG
SDP
Catalog No. 5000453019
Encompass_Preferred
Deoxycytidine triphosphate (dCTP) disodium is a nucleoside triphosphate that contains the pyrimidine base cytosine. It is for research use only and not sold to patients.
  • Solid appearance
  • White to off-white color
  • Stored at -20°C, protected from light, and under nitrogen
  • Soluble in H2O at 125 mg/mL (244.56 mM)
Catalog No. 50-004-53019 Supplier Medchemexpress LLC Supplier No. HYW0107715MG
Deoxycytidine triphosphate (dCTP) disodium is a nucleoside triphosphate that contains the pyrimidine base cytosine. It is for research use only and not sold to patients.
  • Solid appearance
  • White to off-white color
  • Stored at -20°C, protected from light, and under nitrogen
  • Soluble in H2O at 125 mg/mL (244.56 mM)