Learn More
Medchemexpress LLC Deoxycytidine triphosphate trisodium | 2056-98-6 | 98.2% | 467.16 g/mol | C9H16N3O13P3 | 1 ML
SDP
Catalog No. 5000211149
Encompass_Preferred
Deoxycytidine triphosphate is a deoxynucleoside triphosphate used as a substrate for DNA synthesis in PCR, sequencing, and cDNA synthesis. Supplied either as a ready-to-use aqueous stock or as solid material for preparing custom stocks. CAS 2056-98-6; molecular weight 467.16 g/mol; molecular formula C9H16N3O13P3.
  • Ready-to-use 10 mM aqueous stock for rapid assay setup.
  • Solid formats for flexible stock concentration preparation.
  • High purity suitable for molecular biology applications.
  • Soluble in water; example preparation yields 50 mg/mL ≈ 107 mM.
  • Compatible with PCR, DNA sequencing, and cDNA synthesis workflows.
Catalog No. 50-002-11149 Supplier Medchemexpress LLC Supplier No. HY10140010MM 1ML
Deoxycytidine triphosphate is a deoxynucleoside triphosphate used as a substrate for DNA synthesis in PCR, sequencing, and cDNA synthesis. Supplied either as a ready-to-use aqueous stock or as solid material for preparing custom stocks. CAS 2056-98-6; molecular weight 467.16 g/mol; molecular formula C9H16N3O13P3.
  • Ready-to-use 10 mM aqueous stock for rapid assay setup.
  • Solid formats for flexible stock concentration preparation.
  • High purity suitable for molecular biology applications.
  • Soluble in water; example preparation yields 50 mg/mL ≈ 107 mM.
  • Compatible with PCR, DNA sequencing, and cDNA synthesis workflows.