Learn More
TARGETMOL CHEMICALS INC Deoxycytidine triphosphate
SDP
Catalog No. 502876859
Encompass_Preferred
Also available in 10 mg and bulk. Please contact Fisher for quotes. Deoxycytidine triphosphate is a nucleoside triphosphate.It is a raw material in DNA synthesis. Deoxycytidine triphosphate has many applications, such as real-time PCR, cDNA synthesis, and DNA sequencing. Purity 95.19%
Catalog No. 50-287-6859 Supplier TARGETMOL CHEMICALS INC Supplier No. T752050MG
Also available in 10 mg and bulk. Please contact Fisher for quotes. Deoxycytidine triphosphate is a nucleoside triphosphate.It is a raw material in DNA synthesis. Deoxycytidine triphosphate has many applications, such as real-time PCR, cDNA synthesis, and DNA sequencing. Purity 95.19%