Learn More
Medchemexpress LLC Deoxyelephantopin | 29307-03-7 | 5 MG
SDP
Catalog No. 5000421630
Encompass_Preferred
Deoxyelephantopin is a natural bioactive sesquiterpene lactone derived from Elephantopus scaber. It has demonstrated promising anticancer effects across a wide range of cancers by inhibiting NF-κB, MAPK, PI3K/Akt, and β-catenin signaling pathways.
  • Inhibits the cell growth of HCT 116 (colorectal), K562 (chronic myeloid leukemia), KB (oral), and T47D (breast) cancer cell lines.
  • Increases the expression of p53, p-JNK, and p-p38 in cancer cells.
  • Decreases the expression of p-STAT3 and p-mTOR in cancer cells.
  • Downregulates MMP-2, MMP-9, uPA, and uPAR mRNA levels in cancer cells.
Catalog No. 50-004-21630 Supplier Medchemexpress LLC Supplier No. HYN24915MG
Deoxyelephantopin is a natural bioactive sesquiterpene lactone derived from Elephantopus scaber. It has demonstrated promising anticancer effects across a wide range of cancers by inhibiting NF-κB, MAPK, PI3K/Akt, and β-catenin signaling pathways.
  • Inhibits the cell growth of HCT 116 (colorectal), K562 (chronic myeloid leukemia), KB (oral), and T47D (breast) cancer cell lines.
  • Increases the expression of p53, p-JNK, and p-p38 in cancer cells.
  • Decreases the expression of p-STAT3 and p-mTOR in cancer cells.
  • Downregulates MMP-2, MMP-9, uPA, and uPAR mRNA levels in cancer cells.