Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Deoxyepinephrine Hydrochloride, Spectrum™ Chemical
SDP

Catalog No. 18620636 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-636 1 g
18-620-635 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-620-636 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D23611GM
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synonym: N-Methyldopamine Hydrochloride, Ephinine Hydrochloride, Methyldopamine Hydrochloride

Deoxyepinephrine Hydrochloride, also known as N-methyl dopamine and epinine, is an organic compound founds in plants and animals which can be used to treat congestive heart failure. Ungraded products supplied by Spectrum™ are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

N-Methyldopamine Hydrochloride, Ephinine Hydrochloride, Methyldopamine Hydrochloride

Chemical Identifiers

CAS 62-32-8

Specifications

Quantity 1 g
Molecular Weight (g/mol) 203.67
Percent Purity 95%
Grade Ungraded
Physical Form Powder or Waxy Solid
Chemical Name or Material Deoxyepinephrine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.