Learn More
Medchemexpress LLC Deoxyguanosine triphosphate trisodium | 93919-41-6 | 573.13 | 5 ML
SDP
Catalog No. 5000452066
Encompass_Preferred
Deoxyguanosine triphosphate (dGTP) trisodium, 100 mM Solution, PCR Grade is one of the raw materials used by DNA polymerase to synthesize long DNA chains during DNA replication. It is also used in reverse transcription polymerase chain reaction (RT-PCR) for DNA amplification. This product is provided in the form of aqueous solution.
  • Provided in the form of aqueous solution.
  • Used by DNA polymerase to synthesize long DNA chains during DNA replication.
  • Used in reverse transcription polymerase chain reaction (RT-PCR) for DNA amplification.
Catalog No. 50-004-52066 Supplier Medchemexpress LLC Supplier No. HYW008661A5ML
Deoxyguanosine triphosphate (dGTP) trisodium, 100 mM Solution, PCR Grade is one of the raw materials used by DNA polymerase to synthesize long DNA chains during DNA replication. It is also used in reverse transcription polymerase chain reaction (RT-PCR) for DNA amplification. This product is provided in the form of aqueous solution.
  • Provided in the form of aqueous solution.
  • Used by DNA polymerase to synthesize long DNA chains during DNA replication.
  • Used in reverse transcription polymerase chain reaction (RT-PCR) for DNA amplification.