Learn More
Medchemexpress LLC Deoxyguanosine triphosphate trisodium solution | 93919-41-6 | 99.95% | 573.13 | 28.66 MG
SDP
Catalog No. 5000452067
Encompass_Preferred
Deoxyguanosine triphosphate (dGTP) trisodium solution (100mM) is a nucleotide precursor in cells for DNA synthesis. It is used in reverse transcription-polymerase chain reaction (RT-PCR) for DNA amplification. Deoxyguanosine triphosphate is abnormally elevated in the red blood cells of individuals with immunodeficiency related to purine nucleoside phosphorylase deficiency, and it can be regarded as a potential toxic metabolite.
  • Nucleotide precursor for DNA synthesis.
  • Used in reverse transcription-polymerase chain reaction (RT-PCR) for DNA amplification.
Catalog No. 50-004-52067 Supplier Medchemexpress LLC Supplier No. HYW00866128.66MG
Deoxyguanosine triphosphate (dGTP) trisodium solution (100mM) is a nucleotide precursor in cells for DNA synthesis. It is used in reverse transcription-polymerase chain reaction (RT-PCR) for DNA amplification. Deoxyguanosine triphosphate is abnormally elevated in the red blood cells of individuals with immunodeficiency related to purine nucleoside phosphorylase deficiency, and it can be regarded as a potential toxic metabolite.
  • Nucleotide precursor for DNA synthesis.
  • Used in reverse transcription-polymerase chain reaction (RT-PCR) for DNA amplification.