Learn More
Enzo Life Sciences Deoxymannojirimycin . HCl (5 mg). CAS: 73465-43-7
SDP
Supplier:  Enzo Life Sciences BMLS1090005
Encompass_Preferred
Potent inhibitor of α-mannosidase I. Blocks the biosynthesis of complex-type oligosaccharides. Does not inhibit the biosynthesis of lipid-linked oligosaccharides. Induces ER stress in human cells. Purity: ≥98% (TLC). Solubility: Soluble in water, methanol (warm) and DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-1736