Learn More
Enzo Life Sciences Deoxynivalenol (1mg). CAS: 51481-10-8
SDP
Catalog No. 502009641
Encompass_Preferred
Mycotoxin found in cereals. Binds to the ribosome and inhibits protein synthesis. Emetic and refusal activity in pig. Alternative name: DON, 3alpha,7alpha,15-Trihydroxy-12,13-epoxytrichothec-9-en-8-one, Vomitoxin. Purity: ≥98% (TLC, HPLC). Solubility: Soluble in methanol or ethyl acetate; slightly soluble in water. Long Term Storage: +4°C.
Catalog No. 50-200-9641 Supplier Enzo Life Sciences Supplier No. ALX630115M001
Mycotoxin found in cereals. Binds to the ribosome and inhibits protein synthesis. Emetic and refusal activity in pig. Alternative name: DON, 3alpha,7alpha,15-Trihydroxy-12,13-epoxytrichothec-9-en-8-one, Vomitoxin. Purity: ≥98% (TLC, HPLC). Solubility: Soluble in methanol or ethyl acetate; slightly soluble in water. Long Term Storage: +4°C.