Learn More
Medchemexpress LLC HY-N6684 1mg Medchemexpress, Deoxynivalenol CAS:51481-10-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN66841MG
Encompass_Preferred
Medchemexpress, HY-N6684 1mg Deoxynivalenol CAS:51481-10-8 Deoxynivalenol, a mycotoxin of the trichothecenes family, crosses the intestinal mucosa by a paracellular pathway through the tight junctions. The Deoxynivalenol transport is not affected by P-glycoprotein (PgP) or multidrug resistance-associated proteins (MRPs) inhibitors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0454