Learn More
Enzo Life Sciences Deoxynivalenol (5mg). CAS: 51481-10-8
SDP
Catalog No. 502009642
Encompass_Preferred
Mycotoxin found in cereals. Binds to the ribosome and inhibits protein synthesis. Emetic and refusal activity in pig. Alternative name: DON, 3alpha,7alpha,15-Trihydroxy-12,13-epoxytrichothec-9-en-8-one, Vomitoxin. Purity: ≥98% (TLC, HPLC). Solubility: Soluble in methanol or ethyl acetate; slightly soluble in water. Long Term Storage: +4°C.
Catalog No. 50-200-9642 Supplier Enzo Life Sciences Supplier No. ALX630115M005
Add to Cart
Add to Cart
Mycotoxin found in cereals. Binds to the ribosome and inhibits protein synthesis. Emetic and refusal activity in pig. Alternative name: DON, 3alpha,7alpha,15-Trihydroxy-12,13-epoxytrichothec-9-en-8-one, Vomitoxin. Purity: ≥98% (TLC, HPLC). Solubility: Soluble in methanol or ethyl acetate; slightly soluble in water. Long Term Storage: +4°C.