Learn More
AdipoGen Deoxynojirimycin
SDP
Catalog No. 502562093
Encompass_Preferred
Chemical. CAS 19130-96-2. Formula C6H13NO4. MW 163.2. Isolated from Streptomyces sp. Polyhydroxylated piperidine alkaloid. alpha-Glucosidase I and II inhibitor. Antidiabetic. Antihyperglycaemic activity. Animal feedstuff additive. Antiviral. Shows anti-HIV activity.
Catalog No. 50-256-2093 Supplier AdipoGen Supplier No. BVT0112M001
Chemical. CAS 19130-96-2. Formula C6H13NO4. MW 163.2. Isolated from Streptomyces sp. Polyhydroxylated piperidine alkaloid. alpha-Glucosidase I and II inhibitor. Antidiabetic. Antihyperglycaemic activity. Animal feedstuff additive. Antiviral. Shows anti-HIV activity.