Learn More
Cayman Chemical Deoxynojirimycin Tetrabenzyl Ether, Purity: ≥98%, CAS: 69567-11-9, 50 mg
SDP
Catalog No. 502887659
Encompass_Preferred
Starting material for inhibitors of glucosylceramide synthase
Catalog No. 50-288-7659 Supplier Cayman Chemical Supplier No. 1001191450 MG
Starting material for inhibitors of glucosylceramide synthase