Learn More
Cayman Chemical Deoxynojirimycin Tetrabenzyl
SDP
Supplier:  Cayman Chemical 1001191410 MG
Encompass_Preferred
Starting material for inhibitors of glucosylceramide synthase
Catalog No. 50-288-7617