Learn More
New England Biolabs, Inc. Deoxynucleotide (dNTP) Solution Mix – 8 µmol of each
SDP
Catalog No. 50811740
Encompass
Deoxynucleotide (dNTP) Solution Mix is an equimolar solution of ultrapure dATP, dCTP, dGTP and dTTP. A small pack contains 4 vials of 0.2 mls and a large pack contains 5 vials of 0.8 mls. Supplied in: Ultrapure water as a sodium salt at pH 7.5.
  • Compatible with all NEB DNA polymerases in a variety of applications
  • >= 99% pure as determined by HPLC analysis
  • Exceptional stability
Catalog No. 50-811-740 Supplier New England Biolabs, Inc. Supplier No. N0447S
Deoxynucleotide (dNTP) Solution Mix is an equimolar solution of ultrapure dATP, dCTP, dGTP and dTTP. A small pack contains 4 vials of 0.2 mls and a large pack contains 5 vials of 0.8 mls. Supplied in: Ultrapure water as a sodium salt at pH 7.5.
  • Compatible with all NEB DNA polymerases in a variety of applications
  • >= 99% pure as determined by HPLC analysis
  • Exceptional stability