Learn More
New England Biolabs, Inc. Deoxynucleotide (dNTP) Solution Set – 25 µmol of each
SDP
Catalog No. 50811738
Encompass
The Deoxynucleotide (dNTP) Solution Set contains 0.25 ml of each 100 mM ultrapure nucleotide as four separate solutions (dATP, dCTP, dGTP and dTTP). Supplied in: Ultrapure water as a sodium salt at pH 7.5.
Catalog No. 50-811-738 Supplier New England Biolabs, Inc. Supplier No. N0446S
The Deoxynucleotide (dNTP) Solution Set contains 0.25 ml of each 100 mM ultrapure nucleotide as four separate solutions (dATP, dCTP, dGTP and dTTP). Supplied in: Ultrapure water as a sodium salt at pH 7.5.