Learn More
Sigma Aldrich Fine Chemicals Biosciences Deoxynucleoside Triphospha
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 11969064001
Encompass
This product contains 4 separate vials of dATP PCR Grade dCTP PCR Grade dGTP PCR Grade and dTTP PCR Grade each with a concentration of 100 mM and as a clear colorless SOLUTIONon of the sodium salt (pH 8.3).
Catalog No. 50-290-6074