Learn More
Sigma Aldrich Fine Chemicals Biosciences Deoxynucleotide Mix 10 mM
Catalog No. 502914398
Encompass
Nucleotides are organic molecules that serve as the monomers or subunits of nucleic acids (like DNA and RNA). The building blocks of nucleic acids nucleotides consist of a nitrogenous base (purine or pyrimidine) a five-carbon sugar (ribose or deoxyribose) and at least one phosphate group. A nucleoside along with a phosphate group yields a nucleotide.
Catalog No. 50-291-4398 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D729520X.2ML
Nucleotides are organic molecules that serve as the monomers or subunits of nucleic acids (like DNA and RNA). The building blocks of nucleic acids nucleotides consist of a nitrogenous base (purine or pyrimidine) a five-carbon sugar (ribose or deoxyribose) and at least one phosphate group. A nucleoside along with a phosphate group yields a nucleotide.