Learn More
Sigma Aldrich Fine Chemicals Biosciences Deoxynucleotide Set 100 mM
Catalog No. 502905343
Encompass
Deoxynucleotide triphosphates (dNTPs) are highly essential to amplify DNA. This dNTP set contains 100mM SOLUTIONons of individual dNTPs (dATP dCTP dGTP and TTP) in separate tubes for convenience and easy PCR reaction setup.
Catalog No. 50-290-5343 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. DNTP1001KT
Deoxynucleotide triphosphates (dNTPs) are highly essential to amplify DNA. This dNTP set contains 100mM SOLUTIONons of individual dNTPs (dATP dCTP dGTP and TTP) in separate tubes for convenience and easy PCR reaction setup.