Learn More
Cayman Chemical Deoxypodophyllotoxin, Purity: ≥95%, CAS: 19186-35-7, 100 mg
SDP
Catalog No. 502912840
Encompass_Preferred
A flavolignan with diverse biological activities; inhibits tubulin polymerization in a cell-free assay at 50 nM; inhibits COX-2 and 5-LO in isolated mouse bone marrow-derived mast cells stimulated with KL, IL-10, and LPS (IC50s = 1.89 and 0.37 µM, respectively); reduces viral yield in the supernatant of HSV-1- or HSV-2-infected MRC-5 cells at 0.02 µg/ml; inhibits aggregation of rabbit platelets induced by PAF, collagen, or arachidonic acid; induces mortality in P. rapae fifth instar larvae (LC50 = 0.02 g/L); reduces tumor growth in a CT26 murine colon cancer model at 70 µg/kg
Catalog No. 50-291-2840 Supplier Cayman Chemical Supplier No. 36434100 MG
A flavolignan with diverse biological activities; inhibits tubulin polymerization in a cell-free assay at 50 nM; inhibits COX-2 and 5-LO in isolated mouse bone marrow-derived mast cells stimulated with KL, IL-10, and LPS (IC50s = 1.89 and 0.37 µM, respectively); reduces viral yield in the supernatant of HSV-1- or HSV-2-infected MRC-5 cells at 0.02 µg/ml; inhibits aggregation of rabbit platelets induced by PAF, collagen, or arachidonic acid; induces mortality in P. rapae fifth instar larvae (LC50 = 0.02 g/L); reduces tumor growth in a CT26 murine colon cancer model at 70 µg/kg