Learn More
Medchemexpress LLC Deoxypseudouridine | 39967-60-7 | MFCD04973707 | 98.3% | 228.20 | C9H12N2O5 | 5 MG
SDP
Catalog No. 5000247870
Encompass_Preferred
Deoxypseudouridine is a nucleoside analog used in biochemical and molecular biology research as a modified nucleotide building block. Supplied as a white to off-white powder, it is intended for formulation, in vitro assays, and analytical workflows when dissolved in appropriate solvents.
  • Nucleoside analog for nucleotide and nucleic acid research.
  • White to off-white solid, convenient for handling and storage.
  • High purity suitable for analytical and preparative applications.
  • Soluble in DMSO (≈37 mg/mL) with ultrasonic and warming assistance.
  • Stable when stored as powder at -20°C (long-term) or 4°C (short-term).
Catalog No. 50-002-47870 Supplier Medchemexpress LLC Supplier No. HY1019705MG
Deoxypseudouridine is a nucleoside analog used in biochemical and molecular biology research as a modified nucleotide building block. Supplied as a white to off-white powder, it is intended for formulation, in vitro assays, and analytical workflows when dissolved in appropriate solvents.
  • Nucleoside analog for nucleotide and nucleic acid research.
  • White to off-white solid, convenient for handling and storage.
  • High purity suitable for analytical and preparative applications.
  • Soluble in DMSO (≈37 mg/mL) with ultrasonic and warming assistance.
  • Stable when stored as powder at -20°C (long-term) or 4°C (short-term).