Learn More
Worthington Biochemical Corporation DEOXYRIBONLC RIBO PROT 500UN
SDP
Catalog No. NC9748382
Encompass
Molecular Biology Grade. Chromatographically purified to remove RNase and protease. Supplied as a solution at approximately 2 Kunitz units per microliter (approximately 1 mg/ml) containing 50% glycerol and 1 mM calcium chloride.
Catalog No. NC9748382 Supplier Worthington Biochemical Corporation Supplier No. LS006344
Molecular Biology Grade. Chromatographically purified to remove RNase and protease. Supplied as a solution at approximately 2 Kunitz units per microliter (approximately 1 mg/ml) containing 50% glycerol and 1 mM calcium chloride.