Learn More
Worthington Biochemical Corporation DEOXYRIBONUCLEASE 1
SDP
Catalog No. NC9878502
Encompass
Recombinant Bovine pancreatic deoxyribonuclease 1 produced in Pichia pastoris. Chromatographically purified. Free of animal derived components, RNases & proteases. A liquid preparation in 5mM Calcium Acetate, 4mg/ml glycine, pH 5.0 and 50% glycerol.
Catalog No. NC9878502 Supplier Worthington Biochemical Corporation Supplier No. LS006353
Recombinant Bovine pancreatic deoxyribonuclease 1 produced in Pichia pastoris. Chromatographically purified. Free of animal derived components, RNases & proteases. A liquid preparation in 5mM Calcium Acetate, 4mg/ml glycine, pH 5.0 and 50% glycerol.