Learn More
Worthington Biochemical Corporation Deoxyribonuclease I, 125ku
SDP
Catalog No. NC0595335
Encompass
Chromatographically purified. A lyophilized powder containing glycine as a stabilizer. Contains ≤0.0005% RNase. Store at 2-8°C. Protect from moisture. Bovine pancreatic deoxyribonuclease is an endonuclease which splits phosphodiester linkages, preferentially adjacent to a pyrimidine nucleotide yielding polynucleotides with free hydroxyl group at the 3' position and phosphate group at the 5' position. The average chain length of a limit digest is a tetranucleotide.
Catalog No. NC0595335 Supplier Worthington Biochemical Corporation Supplier No. LS006328
Chromatographically purified. A lyophilized powder containing glycine as a stabilizer. Contains ≤0.0005% RNase. Store at 2-8°C. Protect from moisture. Bovine pancreatic deoxyribonuclease is an endonuclease which splits phosphodiester linkages, preferentially adjacent to a pyrimidine nucleotide yielding polynucleotides with free hydroxyl group at the 3' position and phosphate group at the 5' position. The average chain length of a limit digest is a tetranucleotide.