Learn More
Crescent Chemical Co Inc DEOXYRIBONUCLEASE I
SDP
Supplier:  Crescent Chemical Co Inc SE18535.02
Encompass_Preferred
Deoxyribonuclease I from Bovine Pancreas min. 3000 Kunitz units/mg lyophil. Cleaves preferentially double-stranded DNA (in the presence of Mg++-ions single stranded DNA) in oligonucleotides with 5'-terminal phosphate groups. Size - 100MG Storage Conditions - -15 °C TO -25 °C Catalog Number - 18535.0
Catalog No. 50-247-476