Learn More
Worthington Biochemical Corporation Deoxyribonuclease I, 25mg
SDP
Catalog No. NC9131722
Encompass
Partially purified. A lyophilized powder. Store at 2-8°C. Bovine pancreatic deoxyribonuclease is an endonuclease which splits phosphodiester linkages, preferentially adjacent to a pyrimidine nucleotide yielding polynucleotides with free hydroxyl group at the 3' position and phosphate group at the 5' position. The average chain length of a limit digest is a tetranucleotide.
Catalog No. NC9131722 Supplier Worthington Biochemical Corporation Supplier No. LS002138
Partially purified. A lyophilized powder. Store at 2-8°C. Bovine pancreatic deoxyribonuclease is an endonuclease which splits phosphodiester linkages, preferentially adjacent to a pyrimidine nucleotide yielding polynucleotides with free hydroxyl group at the 3' position and phosphate group at the 5' position. The average chain length of a limit digest is a tetranucleotide.