Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Deoxyribonuclease I (Bovine)

Catalog No. 89941796 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-941-796 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-941-796 Supplier Abnova™ Supplier No. P5224
Only null left
Add to Cart
Add to Cart

Used for Enzyme

#REF!

Specifications

Accession Number P00639
For Use With (Application) Enzyme Activity
Gene ID (Entrez) 282217
Molecular Weight (g/mol) 31.3kDa
Name Deoxyribonuclease I (Bovine)
Preparation Method Native protein purified from Bovine Pancreas
Quantity 25 mg
Storage Requirements Store at 4°C.
Regulatory Status RUO
Common Name DNASE1
Gene Symbol DNASE1
Assays Deoxyribonuclease I Assay; Purifying plasmid DNA; Genomic DNA isolation; Molecular weight marker.
Biological Activity ≥2,000 Kunitz units per mg dry weight. One Unit causes an increase in absorbance at 260nm of 0.001 per minute per ml at 25°C when acting upon highly polymerized DNA at pH 5.0.
Species Bovine
Protein Tag None
Expression System Native protein purified from Bovine Pancreas
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.