Learn More
Cusabio Technology LLC deoxyribonuclease I-like 3
Catalog No. 5000050003
Encompass_Preferred
This kit detects DNASE1L3 in human serum plasma tissue homogenates and cell lysates using a quantitative sandwich ELISA method It offers a detection range of 23 44 pg/mL to 1500 pg/mL with a sensitivity of 5 86 pg/mL providing precise measurement for research applications The assay requires 50-100 L of sample per test and delivers results within 1-5 hours Absorbance is measured at 450 nm to determine DNASE1L3 concentration ensuring reliable and reproducible data Designed for flexibility this kit accommodates multiple biological matrices while maintaining high specificity and accuracy in quantifying DNASE1L3 levels across diverse experimental conditions
Catalog No. 50-000-50003 Supplier Cusabio Technology LLC Supplier No. CSBEL007052HU
This kit detects DNASE1L3 in human serum plasma tissue homogenates and cell lysates using a quantitative sandwich ELISA method It offers a detection range of 23 44 pg/mL to 1500 pg/mL with a sensitivity of 5 86 pg/mL providing precise measurement for research applications The assay requires 50-100 L of sample per test and delivers results within 1-5 hours Absorbance is measured at 450 nm to determine DNASE1L3 concentration ensuring reliable and reproducible data Designed for flexibility this kit accommodates multiple biological matrices while maintaining high specificity and accuracy in quantifying DNASE1L3 levels across diverse experimental conditions