Learn More
Medchemexpress LLC Deoxyribonucleic acid from calf thymus | 91080-16-9 | MFCD00163436 | ≥97.0% | N/A | C9H13N3Na3O13P3 | 5 MG
SDP
Catalog No. 5000264911
Encompass_Preferred
Calf thymus DNA is high-quality, double-stranded genomic DNA isolated from calf thymus and provided as a lyophilized powder for molecular biology research. It is suitable as a template or carrier DNA for polymerase assays, hybridization, transfection, and studies of DNA-agent interactions. Store at -20°C and protect from light; in solvent, store at -80°C for up to 6 months.
  • High-quality double-stranded genomic DNA.
  • Suitable substrate for DNA polymerase assays and long-fragment amplification.
  • Acts as carrier or blocking DNA in hybridization and transfection workflows.
  • Available in multiple convenient package sizes.
  • Quality documents provided: datasheet, COA, and SDS.
Catalog No. 50-002-64911 Supplier Medchemexpress LLC Supplier No. HY1095175MG
Add to Cart
Add to Cart
Calf thymus DNA is high-quality, double-stranded genomic DNA isolated from calf thymus and provided as a lyophilized powder for molecular biology research. It is suitable as a template or carrier DNA for polymerase assays, hybridization, transfection, and studies of DNA-agent interactions. Store at -20°C and protect from light; in solvent, store at -80°C for up to 6 months.
  • High-quality double-stranded genomic DNA.
  • Suitable substrate for DNA polymerase assays and long-fragment amplification.
  • Acts as carrier or blocking DNA in hybridization and transfection workflows.
  • Available in multiple convenient package sizes.
  • Quality documents provided: datasheet, COA, and SDS.