Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Deoxyribonucleic acid, MP Biomedicals™

Catalog No. ICN19512950 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19512950 50 mg
ICN19512990 500 mg
ICN19512991 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19512950 Supplier MP Biomedicals, Inc Supplier No. 0219512950
Add to Cart
Add to Cart

Chemical Identifiers

CAS 99675-55-5
Synonym DNA sodium salt, Desoxiribon

Specifications

Color White
Quantity 50 mg
Formula Weight ≈6,000,000
Physical Form Powder
Chemical Name or Material Deoxyribonucleic acid
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.